Learn More
Medchemexpress LLC Levodopa | 59-92-7 | MFCD00002598 | 99.9% | 197.19 g/mol | C9H11NO4 | 10 MG

Supplier: Medchemexpress LLC HYN0304R10MG
L-DOPA (levodopa) analytical standard intended for research and analytical applications. It serves as a reference standard for qualitative and quantitative analyses and method development, and is commonly used in HPLC, GC, and MS workflows.
- Analytical/reference standard suitable for HPLC, GC, and MS
- Purity 99.86%
- Molecular weight 197.19 g/mol
- CAS number 59-92-7
- Offered in small, lab-scale packages for analytical use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.