Learn More
Medchemexpress LLC Levodopa (L-3,4-dihydroxyphenylalanine) | 586971-29-1 | MFCD35325168 | 98.0% | 198.18 g/mol | C8[13C]H11NO4 | 10 MG

Supplier: Medchemexpress LLC HYN0304S210MG
L-DOPA-13C is a carbon-13 isotopically labeled form of L-DOPA (levodopa) supplied as a research-grade stable isotope tracer for use as an internal standard or metabolic tracer in biochemical and pharmacokinetic studies.
- Isotopically labeled with a single 13C atom for tracer and mass spectrometry applications.
- Provided in small milligram quantities suitable for analytical and research use.
- High reported purity of 98.0% supports accurate quantitation and reduced impurity interference.
- Molecular weight approximately 198.18 g/mol, consistent with labeled L-DOPA.
- Storage recommended at 4°C under nitrogen; in solvent, store at -80°C (6 months) or -20°C (1 month) under nitrogen.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.