Learn More
Medchemexpress LLC Levofloxacin N-oxide | 117678-38-3 | MFCD24386420 | >95.0% | 377.37 | C18H20FN3O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1337875MG
Encompass_Preferred
Levofloxacin N-oxide is an analytical reference standard of the levofloxacin metabolite intended for research and analytical applications. Supplied as a dried solid in small mass packs, it is used for chromatographic method development, impurity profiling, and quality control in pharmaceutical analysis. The compound has formula C18H20FN3O5 and molecular weight 377.37; storage recommendations include 4°C protected from light and long-term storage in solvent at -80°C.
  • Provides reference material for chromatographic method development.
  • Suitable for impurity profiling and stability studies.
  • Available in small mass packages for analytical use.
  • Solid form convenient for handling and storage.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-62533