Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Levoketoconazole | 142128-57-2 | 99.4% | C26H28Cl2N4O4 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYB0105B100MG

Encompass_Preferred

Levoketoconazole is the 2S,4R enantiomer derived from racemic Ketoconazole. It is a potent and orally active cortisol synthesis inhibitor. It inhibits key steps in cortisol and testosterone synthesis by inhibiting CYP11B1, CYP11A1, and CYP17A1. It can be used for research on endogenous Cushing's syndrome.

  • Potent and orally active cortisol synthesis inhibitor
  • Inhibits key enzymes: targets CYP11B1, CYP11A1, and CYP17A1
  • Useful for research on endogenous Cushing's syndrome
  • High purity: 99.39% pure
  • Enantiomer specific: 2S,4R enantiomer of Ketoconazole

Catalog No. 50-004-32294


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.