Learn More
Medchemexpress LLC Levoleucovorin (Calcium) | 80433-71-2 | 99.9% | 511.50 | 1 G
SDP
Supplier:  Medchemexpress LLC HY136671G
Encompass_Preferred
Levoleucovorin Calcium (Calcium levofolinate) is the active form of calcium Folinic acid and has anti-tumor effects. It is also a potentiator of the anti-cancer toxicity of 5-Fluorouracil.
  • Anti-tumor effects
  • Potentiator of the anti-cancer toxicity of 5-Fluorouracil
  • High purity
  • Available in solid and solution forms
Catalog No. 50-003-48191