Learn More
Medchemexpress LLC Levomedetomidine hydrochloride | 190000-46-5 | MFCD12912677 | 99.4% | 236.74 | C13H17ClN2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY135735A5MG
Encompass_Preferred
Levomedetomidine hydrochloride is the levorotatory enantiomer of medetomidine, described as a cardioprotective agent with sedative and analgesic effects. Supplied as a white to off-white solid for research use only, it is used in preclinical pharmacology and biochemical studies involving α2-adrenergic receptor activity.
  • Enantiomer of medetomidine with levorotatory configuration.
  • Cardioprotective agent with sedative and analgesic activity.
  • High purity suitable for analytical and preclinical research.
  • Solid, white to off-white physical form for easy handling.
  • Supplied in small pack sizes appropriate for reference studies.
Catalog No. 50-002-22340