Learn More
Selleck Chemical LLC Levomilnacipran Hydrochloride
SDP
Supplier:  Selleck Chemical LLC S569325MG
Encompass_Preferred
Levomilnacipran Hydrochloride is a selective serotonin and norepinephrine reuptake inhibitor with antidepressant activity
Catalog No. 50-000-03703