Learn More
Cayman Chemical LevomIlnacIprnhydrochLRId 10mg
SDP
Supplier:  Cayman Chemical 2395110 MG
Encompass_Preferred
A 5-HT and NE reuptake inhibitor (Kis = 11.2 and 92.2 nM, respectively; IC50s = 19 and 10.5 nM, respectively); selective for 5-HT and NE transporters over DAT (Ki = >10,000 nM) and 23 receptors (Kis = ≥10,000 nM; increases extracellular 5-HT and NE in rat cortex (MEDs = 20 and 2.5 mg/kg); decreases immobility time in the forced swim and tail suspension tests (MEDs = 20 and 2.5 mg/kg, respectively)
Catalog No. 50-299-4422