Learn More
Medchemexpress LLC Levonadifloxacin | 154357-42-3 | 99.8% | 360.38 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14926
Encompass_Preferred
Levonadifloxacin ((S)-(-)-Nadifloxacin; WCK 771) is a broad-spectrum anti-staphylococcal agent that demonstrates antibacterial activity against Methicillin-susceptible *Staphylococcus aureus* (MSSA) and Methicillin-resistant *S. aureus* (MRSA) strains, reducing those phagocytized in THP-1 monocytes. It also shows efficacy against *Staphylococcus aureus* in a Neutropenic Murine Lung Infection Model.
  • Broad-spectrum anti-staphylococcal agent
  • Active against MSSA and MRSA strains
  • Reduces phagocytized bacteria in THP-1 monocytes
  • Efficacious in Neutropenic Murine Lung Infection Model
Catalog No. 50-003-60405