Learn More
Medchemexpress LLC Levonadifloxacin | 154357-42-3 | 99.8% | 360.38 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY149265MG
Encompass_Preferred
Levonadifloxacin, also known as (S)-(-)-Nadifloxacin or WCK 771, is an antibiotic and a broad-spectrum anti-staphylococcal agent. It demonstrates antibacterial activity against both Methicillin-susceptible *Staphylococcus aureus* (MSSA) and Methicillin-resistant *S. aureus* (MRSA) strains, leading to a reduction of those phagocytized in THP-1 monocytes.
  • Broad-spectrum anti-staphylococcal agent
  • Active against MSSA and MRSA strains
  • Reduces phagocytized *S. aureus* in THP-1 monocytes
  • Efficacious in vivo against *Staphylococcus aureus* in a Neutropenic Murine Lung Infection Model
Catalog No. 50-003-60967