Learn More
Medchemexpress LLC Levonorgestrel | 797-63-7 | 99.98% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0257500MG
Encompass_Preferred
Levonorgestrel is an orally active inhibitor of progesterone. It exhibits anticancer activity and can induce apoptosis. This compound serves as a contraceptive and can be utilized in combination with other medications. Additionally, Levonorgestrel is employed in the study of osteoporosis and uterine leiomyoma.
  • Inhibits cell proliferation and promotes apoptosis in uterine leiomyoma cells.
  • Can inhibit progesterone production in luteal cells at high doses.
  • Inhibits bone resorption, decreases bone turnover, and increases bone mineral content in Sprague-Dawley rats.
  • Provides effective contraception in Apodemus agrarius mice when combined with quinestrol.
Catalog No. 50-004-06972