Learn More
Cayman Chemical LevonrdefrIn 10mg
SDP
Supplier:  Cayman Chemical 2399310 MG
Encompass_Preferred
A vasoconstrictor; (–)-enantiomer of nordefrin; rapidly increases blood pressure and heart rate in anesthetized dogs (1-5 UG/kg); in combination with mepivacaine, increases systolic arterial blood pressure in anesthetized dogs (0.514 and 1.542 mg/kg)
Catalog No. 50-293-3225