Learn More
Medchemexpress LLC Levophacetoperane hydrochloride | 23257-56-9 | 98.0% | 269.77 | C14H20ClNO2 | 1 MG

Supplier: Medchemexpress LLC HY1016311MG
Levophacetoperane hydrochloride is the hydrochloride salt of levophacetoperane, a psychostimulant research compound that competitively inhibits norepinephrine and dopamine uptake in vitro. It has historical use as an antidepressant and anorectic and is supplied as a white to off-white solid for laboratory research (not for human use).
- Purity: 98.0%.
- Molecular weight: 269.77.
- Chemical formula: C14H20ClNO2.
- Appearance: white to off-white solid.
- Mechanism: competitive inhibition of norepinephrine and dopamine uptake in vitro.
- Intended use: for research use only, not for human or clinical use.
- Packaging: available in small milligram quantities suitable for analytical and assay work.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.