Learn More
Medchemexpress LLC Levophacetoperane (hydrochloride) | 23257-56-9 | 98.0% | 269.77 | C14H20ClNO2 | 10 MG

Supplier: Medchemexpress LLC HY10163110MG
Levophacetoperane hydrochloride is the hydrochloride salt of levophacetoperane, a psychostimulant research chemical that competitively inhibits norepinephrine and dopamine uptake in vitro. Supplied as a white to off-white solid for laboratory research; not for human or veterinary use.
- Inhibits norepinephrine and dopamine uptake in vitro.
- White to off-white solid form.
- CAS number 23257-56-9.
- Molecular weight 269.77.
- Purity 98.0%.
- Available in small research quantities including 10 MG and DMSO solution.
- Store sealed at 4°C; in solution store at -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.