Learn More
Medchemexpress LLC Levophacetoperane hydrochloride | 23257-56-9 | 98.0% | 269.77 g/mol | C14H20ClNO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1016315MG
Encompass_Preferred
Levophacetoperane hydrochloride is a research-grade compound that competitively inhibits norepinephrine and dopamine uptake in vitro. It is supplied as a white to off-white solid with a reported purity of 98.0% and a molecular weight of 269.77 g/mol. Intended for laboratory research use only; not for human or clinical applications.
  • White to off-white solid for easy handling.
  • Reported purity 98.0%.
  • Inhibits norepinephrine and dopamine uptake in vitro.
  • Available as small solid quantities and as a DMSO solution for flexible use.
  • Datasheet and certificate of analysis available for quality verification.
Catalog No. 50-002-47367