Learn More
Selleck Chemical LLC Levosimendan 50mg 141505-33-1
SDP
Supplier:  Selleck Chemical LLC S244650MG
Encompass_Preferred
Levosimendan is a calcium sensitizer acting through calcium-dependent binding to cardiac troponin C (cTnC), provides treatment for heart failure. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2405