Learn More
Apexbio Technology LLC Levosimendan, 50mg. Cas: 141505-33-1 MFCD: MFCD00867135. calcium sensitiser
SDP
Supplier:  Apexbio Technology LLC B196150
Encompass_Preferred
Levosimendan is a calcium sensitizer acting through calcium-dependent binding to cardiac troponin C (cTnC), provides treatment for heart failure. Phase 4. 141505-33-1. MFCD00867135
Catalog No. 50-101-1569