Learn More
Apexbio Technology LLC Levothyroxine sodium 55-03-8 100mg
SDP
Supplier:  Apexbio Technology LLC C6460100
Encompass_Preferred
Levothyroxine sodium (55-03-8) is a synthetic thyroid hormone analogue It is designed to restore or modulate thyroid hormone levels thereby regulating metabolic and developmental processes Levothyroxine sodium exerts its biological activity primarily through enzymatic conversion to the active form triiodothyronine (T3) by deiodinases after which T3 interacts with nuclear thyroid hormone receptors to modulate gene expression and cellular metabolism Based on these pharmacological properties levothyroxine sodium holds research potential in studies of thyroid dysfunction particularly hypothyroidism and in investigations of thyroid-regulated physiological responses and endocrine interactions
Catalog No. 50-000-90088