Learn More
Medchemexpress LLC Lewis a trisaccharide | 56570-03-7 | 529.49 | 1 MG

Supplier: Medchemexpress LLC HYN105311MG
Lewis a trisaccharide (Lewis a) is a trisaccharide that has been found to be present in the glycan structures of spermatozoa. Lewis a trisaccharide also is a major component of the glycan structures on the surface of HL-60 cells.
- Trisaccharide
- Present in the glycan structures of spermatozoa
- Major component of glycan structures on the surface of HL-60 cells
- Purity: 95.0%
- Appearance: Solid
- Color: White to off-white
- Initial source: Animals, human milk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.