Learn More
Selleck Chemical LLC Lexibulin (CYT997) S2195-5mg
SDP
Supplier:  Selleck Chemical LLC S21955MG
Encompass_Preferred
Lexibulin (CYT997 SRI-32007) is a potent microtubule polymerization inhibitor with IC50 of 10-100 nM in cancer cell lines Phase 2
Catalog No. 50-310-8119