Learn More
SOLVENTUM 3M LifeASSURE PDA Series Filter Cartridge | 20in | 0.2um ABS | Sanitary | 1/Case
Supplier:  SOLVENTUM 7000051530
Encompass_Preferred
3M combines two asymmetric polyethersulfone (PES) membrane layers with Advanced Pleat Technology in manufacturing LifeASSURE PDA Series Filter Cartridges, creating a filter that provides high throughput and fast flow. These filters have applications in pre-filtration and sterilizing filtration in the pharmaceutical, biological, and bioprocess industries where low protein binding is critical
Catalog No. NC1693864