Learn More
Enzo Life Sciences LFM-A13 (25mg). CAS: 62004-35-7, 244240-24-2
SDP
Supplier:  Enzo Life Sciences BMLEI2950025
Encompass_Preferred
Bruton's tyrosine kinase (BTK) inhibitor IC50= 2.5 µM. IC50's for JAK-1, JAK-2, JAK-3, SYK, HCK, EGFR kinase, IR kinase all >300 µM Purity: ≥97% (TLC). Solubility: Soluble in DMSO (10mg/ml) or ethanol (0.3mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0678