Learn More
Medchemexpress LLC LGD 3303 | 917891-35-1 | ≥98.0% | 342.7 g/mol | C16H14ClF3N2O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10357625MG
Encompass_Preferred
LGD 3303 is a non-steroidal selective androgen receptor modulator (SARM) used in preclinical research as a potent androgen receptor agonist. It selectively binds the human androgen receptor (reported Ki ≈ 0.9 nM) and has demonstrated anabolic effects on muscle mass and bone density in animal studies.
  • Non-steroidal SARM for research use only
  • High affinity for the androgen receptor (Ki ≈ 0.9 nM)
  • Shows anabolic effects on muscle and bone in animal models
  • Offered as a high-purity reference standard (≥98%)
  • Typical storage at -20 °C to preserve stability
Catalog No. 50-002-44966