Learn More
TARGETMOL CHEMICALS INC LGD-6972 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T118455MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. LGD-6972 is an antagonist of glucagon receptor. Purity 99.33%
Catalog No. 50-287-5907