Learn More
Cayman Chemical LGD 6972amonIum salt 5mg
SDP
Supplier:  Cayman Chemical 409615 MG
Encompass_Preferred
A GCGR antagonist; reduces glucagon-induced hyperglycemia in wild-type mice; reduces blood glucose levels in mouse models of diabetes
Catalog No. 50-294-5077