Learn More
Selleck Chemical LLC LGK-974
SDP
Supplier:  Selleck Chemical LLC S714310MM/1ML
Encompass_Preferred
LGK-974 (NVP-LGK974 WNT974) is a potent and specific PORCN inhibitor and inhibits Wnt signaling with IC50 of 0 4 nM in TM3 cells Phase 1
Catalog No. 50-000-01324