Learn More
Medchemexpress LLC LHC-165 | 1258595-14-0 | 98.0% | 603.55 | 10 MG

Supplier: Medchemexpress LLC 10MGHY111786
LHC-165 is a TLR7 agonist with potential for use in the study of solid tumors. It is for research use only and not sold to patients.
- Target: TLR7
- Molecular weight: 603.55
- Formula: C29H32F2N3O7P
- Appearance: Solid
- Color: Light yellow to yellow
- Solubility (in vitro): 1M NaOH: 50 mg/mL (82.84 mM; ultrasonic and adjust pH to 12 with NaOH)
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: -20°C, stored under nitrogen. In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)
- Disease areas: Cancer, Cancer immunotherapy
- Immunology/inflammation: Toll-like receptor (TLR)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.