Learn More
TARGETMOL CHEMICALS INC LHF-535 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T11138L25MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. LHF-535 is an antiviral compound. It has EC50s of <1 uM, <1 uM, <1 uM, and 1-10 uM for Lassa, Machupo, Junin, and VSVg virus, respectively. Purity 99.9%
Catalog No. 50-288-0783