Learn More
Medchemexpress LLC LI-2242 | 2762762-17-2 | C20H18Cl2N2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15353050MG
Encompass_Preferred
LI-2242 is an inositol hexakisphosphate kinase (IP6K) inhibitor, demonstrating potent inhibition of IP6K1, IP6K2, IP6K3, and IPMK. It is suitable for research in areas such as type II diabetes, obesity, metabolic complications, venous thrombosis, and psychiatric disorders.
  • Inhibits IP6K1 with an IC50 of 31 nM
  • Inhibits IP6K2 with an IC50 of 42 nM
  • Inhibits IP6K3 with an IC50 of 8.7 nM
  • Inhibits IPMK with an IC50 of 1944 nM
  • Useful for research on type II diabetes, obesity, and metabolic complications
  • Applicable for studies on venous thrombosis and psychiatric disorders
Catalog No. 50-003-73018