Learn More
Selleck Chemical LLC Liarozole dihydrochloride
SDP
Supplier:  Selleck Chemical LLC S659425MG
Encompass_Preferred
Liarozole Dihydrochloride is identified as a modest inhibitor of P450 with IC50 of 2 2 6 0 M and 260 nM for CYP26 and rat CYP17 respectively
Catalog No. 50-000-02467