Learn More
Medchemexpress LLC Libvatrep | 871814-52-7 | 99.5% | 305.33 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY114017100MG
Encompass_Preferred
Libvatrep is a potent vanilloid receptor TRPV1 antagonist. Extracted from patent US8349852B2, compound B8, this chemical is suitable for various research applications.
  • Potent vanilloid receptor TRPV1 antagonist
  • Suitable for research applications
  • Available in solid form
Catalog No. 50-004-37820