Learn More
Medchemexpress LLC Libvatrep | 871814-52-7 | MFCD18711490 | 99.5% | 305.33 | C18H15N3O2 | 5 MG

Supplier: Medchemexpress LLC HY1140175MG
Libvatrep is a research-grade TRPV1 (vanilloid receptor) antagonist supplied as a solid for in vitro and in vivo pharmacology studies. The compound is provided with analytical documentation and recommended solvent/formulation protocols for experimental use, and is identified by CAS 871814-52-7.
- Potent TRPV1 antagonist for pharmacological research.
- High purity (~99.5%) suitable for analytical assays.
- Molecular weight 305.33 and formula C18H15N3O2.
- Available as small solid quantities and as DMSO solution formats.
- Formulation guidance provided for in vitro and in vivo use.
- Certificate of analysis and safety data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.