Learn More
Abcam Licarbazepine (10,11-Dihydro-10-hydro x ycarbamazepine), Active o x carbazepine metabolite, 50MG
Supplier:  Abcam AB14586750MG
Encompass_Preferred
MW 254.28 Da, Purity >99%. Active metabolite of oxcarbazepine (ab141827). Induces microsomal enzymes via sodium channel protein inhibition. Potentiates minor GABAA receptor currents; exhibits anticonvulsant activity. Antiepileptic agent. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5058