Learn More
LiChropur™ Hydrogen Chloride Solution, 0.5 M in Methanol, For GC Derivatization, MilliporeSigma™ Supelco™
$151.20 - $189.50
Specifications
| MDL Number | MFCD00011324 |
|---|---|
| Density | 0.800 to 0.811 g/mL (at 20°C) |
| Synonym | Methanolic HCl |
| Formula Weight | 36.46 |
| Grade | GC Derivatization |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
11-102-0192
|
MilliporeSigma Supelco
0760710X1ML |
10 x 1 mL |
Each for $151.20
|
|
|||||
|
11-101-9776
|
MilliporeSigma Supelco
07607100ML |
100 mL |
Each for $189.50
|
|
|||||
Description
Specifications
| MFCD00011324 | |
| Methanolic HCl | |
| GC Derivatization |
| 0.800 to 0.811 g/mL (at 20°C) | |
| 36.46 |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.