Learn More
LiChropur™ Hydrogen Chloride Solution, 0.5 M in Methanol, For GC Derivatization, MilliporeSigma™ Supelco™
Supplier:  MilliporeSigma Supelco 07607100ML
 View more versions of this product
Catalog No. 11-101-9776
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7647-01-0
MFCD00011324
Methanolic HCl
GC Derivatization
HCl
0.800 to 0.811 g/mL (at 20°C)
36.46
100 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Product Title
Select an issue
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany