Learn More
LiChropur™ Hydrogen Chloride Solution, 3 M in Methanol, For GC Derivatization, MilliporeSigma™ Supelco™
Supplier:  MilliporeSigma Supelco 9096410X1ML
 View more versions of this product
Catalog No. 11-102-0141
Add to Cart
Description
Description
Methanolic HCl (hydrogen chloride in methanol) is particularly useful for preparing methyl esters of volatile (short chain) fatty acids. Fatty acids are esterified by heating them with an anhydrous alcohol (e.g., methanol) in the presence of an acidic catalyst (e.g., HCl) in a sealed vessel at a high temperature for a short time. In the reaction, a fatty acid molecule and an alcohol molecule are joined, with the release of a water molecule. The derivatives can be quickly and easily recovered, quantitatively, from the reaction medium.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7647-01-0
MFCD00011324
Methanolic HCl
10 x 1 mL
HCl
0.871 to 0.881 g/mL (at 20°C)
GC Derivatization
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Product Title
Select an issue
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany