Learn More
Medchemexpress LLC Licochalcone D | 144506-15-0 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN41871MG
Encompass_Preferred
Licochalcone D, a flavonoid compound mainly existing in the root of Glycyrrhiza uralensis, is a potent and orally active inhibitor of NF-kappaB (NF-κB) p65. Licochalcone D possesses antioxidant, anti-inflammatory, and anti-cancer properties.
  • Potent and orally active inhibitor of NF-kappaB (NF-κB) p65
  • Exhibits antioxidant, anti-inflammatory, and anti-cancer properties
  • Negatively regulates LPS-induced NF-κB activation
  • Inhibits LPS-induced expression of TNFα and MCP-1
  • Inhibits LPS-induced iNOS expression and NO production
  • Inhibits the proliferation of human melanoma cells
  • Induces apoptosis of A375 cells
  • Decreases mitochondrial membrane potential and increases ROS production
  • Decreases the migration and invasion ability of A375 cells
  • Inhibits tumor growth in a mouse xenograft model of murine melanoma cells
Catalog No. 50-004-20433