Learn More
Medchemexpress LLC Licochalcone E | 864232-34-8 | 1 MG
Supplier: Medchemexpress LLC HYN41821MG
Licochalcone E is a flavonoid compound isolated from *Glycyrrhiza uralensis*. It inhibits NF-κB and AP-1 transcriptional activity by inhibiting AKT and MAPK activation. This compound is intended for research use only.
- Isolated from Glycyrrhiza uralensis.
- Inhibits NF-κB and AP-1 transcriptional activity.
- Inhibits AKT and MAPK activation.
- Targets Akt, p38 MAPK, and Autophagy.
- Involved in PI3K/Akt/mTOR and MAPK/ERK pathways.