Learn More
Medchemexpress LLC HY-N4185 5mg Medchemexpress, Licoflavone A CAS:61153-77-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN41855MG
Encompass_Preferred
Medchemexpress, HY-N4185 5mg Licoflavone A CAS:61153-77-3 Licoflavone A is a flavonoid isolated from the roots of Glycyrrhiza uralensis, inhibits protein tyrosine phosphatase-1B (PTP1B), with an IC50 of 54.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0433