Learn More
Medchemexpress LLC Licogliflozin | 1291094-73-9 | 98.0% | 416.46 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10909250MG
Encompass_Preferred
Licogliflozin is a sodium glucose cotransporter (SGLT1 and SGLT2) inhibitor. It is also described as a non-anti-fibrotic treatment agent for non-alcoholic steatohepatitis (NASH).
  • Sodium glucose cotransporter (SGLT1 and SGLT2) inhibitor.
  • Non-anti-fibrotic treatment agent for non-alcoholic steatohepatitis (NASH).
Catalog No. 50-003-44499