Learn More
Medchemexpress LLC HY-N3389 5mg Medchemexpress, Licoisoflavone A CAS:66056-19-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN33895MG
Encompass_Preferred
Medchemexpress, HY-N3389 5mg Licoisoflavone A CAS:66056-19-7 Licoisoflavone A, an isoflavone, mainly derived from Glycyrrhiza uralensis Fisch.. Licoisoflavone A inhibits lipid peroxidation with an IC50 of 7.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0412