Learn More
Medchemexpress LLC Licoisoflavone A | 66056-19-7 | MFCD04039831 | 99.4% | 354.35 | C20H18O6 | 10 MG

Supplier: Medchemexpress LLC HYN338910MG
Licoisoflavone A is an isoflavone research compound supplied as a powder for biochemical and pharmacological studies. It has reported antiproliferative and pro-apoptotic activity in colorectal cancer models and shows dose-dependent inhibition of SARS-CoV-2 in cell-based assays.
- High purity (99.4%) suitable for research use.
- Provided as a 10 MG powder for laboratory assays.
- CAS number 66056-19-7; chemical formula C20H18O6; molecular weight 354.35.
- Exhibits inhibition of CDK2-Cyclin E1 and lipid peroxidation (IC50 = 7.2 μM).
- Storage: powder -20°C (long term) or 4°C (short term).
- Intended for in vitro and in vivo studies of colorectal cancer and viral infection models.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.