Learn More
TARGETMOL CHEMICALS INC LICOSTINEL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T278285MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Licostinel (CGP 63446) is a glutamate receptor antagonist. Licostinel protects against permanent focal cerebral ischemia. purity: 98%
Catalog No. 50-258-0569