Learn More
Uline Lid for Bulk Containers H-2093 and H-4051 - 32 x 30 | | 1 EA
Supplier:  Uline H4101
Encompass
Lids keep contents free from dust and debris during transit or storage; Raised edges keep containers secure when stacked; Lids rests around rim of container; Compatible Collapsible Bulk Containers: 32 x 30 x 25, 32 x 30 x 34; Adds 1 1/4 to height of Bulk Container; MATERIAL: HDPE High Density Polyethylene; TEMPERATURE: 0 - 120 F; USAGE: Lid rests around rim of container does not lock or snap on; When stacking on top of lid, drop doors and side panels must be upright and fully locked in place
Catalog No. NC2469043