Learn More
XRF Scientific Americas Inc. PLATINUM LID FOR STANDARD SHAPE CRUCIBLE 10ML FROM XRF SCIENTIFIC AMERICAS
Supplier:  XRF Scientific Americas Inc. GCS10LID
Encompass
Lid suitable for a 10 ml Standard shape crucible. These 99.95% pure platinum lids are used for Fusions, Digestion, Ash testing, Loss on Ignition (LOI). Due to their high temperature strenght, ductility, corrosion and oxidation, excellent resistance to chemical attacks by acids and fusion mixtures resistance; these platinum crucibles are critical when high temperatures & resistance to chemical attacks are required.
Catalog No. NC1405007