Learn More
Research Products International Corp Lid for Staining Dish, 20x30
SDP
Supplier:  Research Products International Corp 248932L
Encompass
Ideal for all staining, fixing and destaining procedures. Heavy duty staining dishes are molded from clear Pyrex brand borosilicate glass. Offers best compatibility with all chemical buffers and reagents. Safe to use with electrophoresis stains and dyes. Clean thoroughly after use. Supplied with tight fitting polyethylene lids to prevent evaporation of solutions.
Catalog No. 50-197-8693