Learn More
Medchemexpress LLC Lidocaine-d10 | 851528-09-1 | 99.58% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0185S11MG
Encompass_Preferred
Lidocaine-d10 is deuterium-labeled Lidocaine, an amide derivative that inhibits sodium channels. It decreases the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating MEK/ERK and NF-κB signaling pathways. It has potential for the research of ventricular arrhythmia.
Catalog No. 50-004-05766