Learn More
U.S. Pharmacopeia Lidocaine hydrochloride, 6108-05-0, MFCD00150329, 150 mg
Supplier:  U.S. Pharmacopeia 1366013
Encompass
Empirical Formula (Hill Notation): C14H22N2O · HCl · H2O, Molecular Weight: 288.81, Synonym: 2-Diethylamino-N-(2,6-dimethylphenyl)acetamide hydrochloride monohydrate, Lidocaine hydrochloride monohydrate, Lignocaine hydrochloride monohydrate, Xylocaine hydrochloride monohydrate.
Catalog No. NC1356705