Learn More
Selleck Chemical LLC Lidocaine hydrochloride S4667-500mg
SDP
Supplier:  Selleck Chemical LLC S4667500MG
Encompass_Preferred
Lidocaine hydrochloride (Lidothesin Lignocaine Xyloneural) is an inhibitor of epidermal growth factor receptor (EGFR) and has antiproliferative effect on human tongue cancer cells Lidocaine hydrochloride is a local anesthetic and cardiac depressant used as an antiarrhythmia agent
Catalog No. 50-311-2158