Learn More
Medchemexpress LLC Lidocaine hydrochloride | 73-78-9 | 99.84% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0185A10MM/1ML
Encompass_Preferred
Lidocaine hydrochloride, also known as Lignocaine hydrochloride, is an amide derivative that inhibits sodium channels with complex voltage and use dependence. It has been shown to decrease the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating MEK/ERK and NF-κB signaling pathways. This compound is used to treat ventricular arrhythmia and acts as an effective tumor-inhibitor. For research use only.
  • Inhibits sodium channels
  • Decreases gastric carcinoma cell proliferation and migration
  • Up-regulates miR-145 expression
  • Inactivates MEK/ERK and NF-κB signaling pathways
  • Used for ventricular arrhythmia treatment
  • Acts as an effective tumor-inhibitor
  • Causes reversible tail nerve block in rats
  • Purity: 99.84%
  • Solid appearance
Catalog No. 50-004-06635